Assay Details
| Gene Symbol: DLGAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005258174.4 | XP_005258231.1 | 1 - 2 | 370 | 69 | |
| XM_017026083.1 | XP_016881572.1 | 1 - 2 | 369 | 69 | ||
| XM_017026084.1 | XP_016881573.1 | 1 - 2 | 366 | 69 | ||
More Information
Set Membership: |
|

DLGAP1
Genomic Map