Assay Details
| Gene Symbol: CYSLTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282186.1 | NP_001269115.1 | 2 | 2499 | 116 | |
| NM_001282187.1 | NP_001269116.1 | 4 | 2669 | 116 | ||
| NM_001282188.1 | NP_001269117.1 | 4 | 2646 | 116 | ||
| NM_006639.3 | NP_006630.1 | 3 | 2586 | 116 | ||
| GenBank mRNA | DQ131803.1 | - | 1 | 1143 | 116 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TATA-box binding protein associated factor 9b | TAF9B |
DN-7, DN7, TAF9L, TAFII31L, TFIID-31 |
-131.8kb | Sense | Chr. X - 78129748 - 78139682 on NCBI Build GRCh38 |
| Downstream | zinc finger CCHC-type containing 5 | ZCCHC5 |
Mar3, Mart3, ZHC5 |
382.3kb | Sense | Chr. X - 78656069 - 78659328 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CYSLTR1
Genomic Map