Assay Details
| Gene Symbol: USE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018467.3 | NP_060937.1 | 4 - 5 | 446 | 67 | |
| XM_017026975.1 | XP_016882464.1 | 5 - 6 | 1156 | 67 | ||
| XM_017026976.1 | XP_016882465.1 | 3 - 4 | 396 | 67 | ||
| GenBank mRNA | AB097050.1 | - | 4 - 5 | 417 | 67 | |
| AF220052.1 | - | 4 - 5 | 417 | 67 | ||
| BC008455.1 | - | 4 - 5 | 394 | 4247241 | 67 | |
| BF448659.1 | - | 4 - 5 | 67 | |||
| BI758045.1 | - | 4 - 5 | 433 | 67 | ||
More Information
Set Membership: |
|

USE1
Genomic Map