Assay Details
| Gene Symbol: NFATC2IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032815.3 | NP_116204.3 | 2 - 3 | 482 | 94 | |
| GenBank mRNA | AF458593.1 | - | 2 - 3 | 492 | 94 | |
| AK074761.1 | - | 2 - 3 | 472 | 94 | ||
| BC080628.1 | - | 2 - 3 | 149 | 94 | ||
| BC101741.1 | - | 2 - 3 | 542 | 8069247 | 94 | |
| BC112182.1 | - | 2 - 3 | 542 | 8327650 | 94 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000313.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16004414.hg.1 | ENST00000320805.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16004414.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16004414.hg.1 | TC16000313.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000562977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000562977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000562977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000562977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000562977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000564978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000564978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000564978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000564978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000564978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000564978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000564978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000564978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000564978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000565752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000565752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000565752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000565752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000565919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000565919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000565919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000565919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000568998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000568998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000568998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000568998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | ENST00000578855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007374.hg.1 | NR_039742.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600153239.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153239.hg.1 | ENST00000568998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153239.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600153239.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153239.hg.1 | ENST00000568998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153239.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600153239.hg.1 | TC1600007374.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600153238.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153238.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600153238.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600153238.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600153238.hg.1 | TC1600007374.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000320805.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000562977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000562977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000562977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000562977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000562977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000564978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000564978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000564978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000564978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000564978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000564978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000564978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000564978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000564978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000565752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000565752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000565752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000565752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000565919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000565919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000565919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000565919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000568148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000568998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000568998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000568998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000568998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | ENST00000578855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | NM_032815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007374.hg.1 | NR_039742.1 | Refseq |

NFATC2IP
Genomic Map