Assay Details
| Gene Symbol: GPRC5D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005253421.3 | XP_005253478.1 | 2 - 3 | 2394 | 69 | |
| XM_017019582.1 | XP_016875071.1 | 1 - 2 | 487 | 69 | ||
| XM_017019583.1 | XP_016875072.1 | 1 - 2 | 486 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005253421.3, XM_017019582.1, XM_017019583.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000394742.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000394742.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000394742.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000538231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000538231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000542078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000542078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000542078.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | NR_038920.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | NR_038920.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | NR_038920.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | NR_038920.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | NR_038920.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | NR_038920.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | NR_038920.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | NR_038920.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006898.hg.1 | NR_038920.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009980.hg.1 | ENST00000228887.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009980.hg.1 | ENST00000228887.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009980.hg.1 | ENST00000228887.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009980.hg.1 | ENST00000228887.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009980.hg.1 | ENST00000396333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009980.hg.1 | ENST00000396333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009980.hg.1 | ENST00000541128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009980.hg.1 | ENST00000541128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009980.hg.1 | ENST00000541128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009980.hg.1 | NM_018654.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009980.hg.1 | NM_018654.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009980.hg.1 | NM_018654.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009980.hg.1 | NM_018654.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000394742.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000394742.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000394742.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000499948.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000536029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000538231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000538231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000540198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000542078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000542078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000542078.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000543515.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000545914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | ENST00000607894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | NR_038920.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | NR_038920.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | NR_038920.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | NR_038920.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | NR_038920.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | NR_038920.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | NR_038920.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | NR_038920.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006898.hg.1 | NR_038920.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009980.hg.1 | ENST00000228887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009980.hg.1 | ENST00000228887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009980.hg.1 | ENST00000228887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009980.hg.1 | ENST00000228887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009980.hg.1 | ENST00000396333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009980.hg.1 | ENST00000396333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009980.hg.1 | ENST00000541128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009980.hg.1 | ENST00000541128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009980.hg.1 | ENST00000541128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009980.hg.1 | NM_018654.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009980.hg.1 | NM_018654.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009980.hg.1 | NM_018654.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009980.hg.1 | NM_018654.1 | Refseq |

GPRC5D
Genomic Map