Assay Details
| Gene Symbol: AQP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170.2 | NP_001161.1 | 6 - 7 | 860 | 83 | |
| NM_001318156.1 | NP_001305085.1 | 5 - 6 | 742 | 83 | ||
| NM_001318157.1 | NP_001305086.1 | 6 - 7 | 1178 | 83 | ||
| NM_001318158.1 | NP_001305087.1 | 6 - 7 | 860 | 83 | ||
| NR_134514.1 | 5 - 6 | 736 | 83 | |||
| NR_134515.1 | 6 - 7 | 887 | 83 | |||
| XM_005251453.3 | XP_005251510.1 | 7 - 8 | 960 | 83 | ||
| XM_006716765.3 | XP_006716828.1 | 5 - 6 | 621 | 83 | ||
| XM_011517866.2 | XP_011516168.1 | 6 - 7 | 770 | 83 | ||
| XM_011517867.2 | XP_011516169.1 | 4 - 5 | 400 | 83 | ||
| XM_017014700.1 | XP_016870189.1 | 6 - 7 | 1061 | 83 | ||
| XM_017014701.1 | XP_016870190.1 | 5 - 6 | 731 | 83 | ||
| XM_017014702.1 | XP_016870191.1 | 6 - 7 | 766 | 83 | ||
| XM_017014703.1 | XP_016870192.1 | 5 - 6 | 519 | 83 | ||
| XM_017014704.1 | XP_016870193.1 | 4 - 5 | 827 | 83 | ||
| XM_017014705.1 | XP_016870194.1 | 4 - 5 | 497 | 83 | ||
| XM_017014706.1 | XP_016870195.1 | 7 - 8 | 891 | 83 | ||
| GenBank mRNA | AB006190.1 | - | 6 - 7 | 697 | 83 | |
| AK294028.1 | - | 6 - 7 | 671 | 83 | ||
| AK297860.1 | - | 6 - 7 | 765 | 83 | ||
| AK297885.1 | - | 6 - 7 | 994 | 83 | ||
| AK300716.1 | - | 3 - 4 | 638 | 83 | ||
| AK310456.1 | - | 4 - 5 | 599 | 83 | ||
| AK315879.1 | - | 3 - 4 | 629 | 83 | ||
| BC062701.1 | - | 5 - 6 | 569 | 30324263 | 83 | |
| BC119672.1 | - | 5 - 6 | 568 | 83 | ||
| BC119673.1 | - | 5 - 6 | 568 | 83 | ||
| DQ786216.1 | - | 1 - 2 | 72 | 83 | ||
More Information
Set Membership: |
|

AQP7
Genomic Map