Assay Details
| Gene Symbol: AK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016282.3 | NP_057366.2 | 1 - 2 | 376 | 62 | |
| GenBank mRNA | AF086913.1 | - | 1 - 2 | 146 | 62 | |
| AF100785.1 | - | 1 - 2 | 154 | 62 | ||
| AK001553.1 | - | 1 - 2 | 294 | 62 | ||
| AK001951.1 | - | 1 - 2 | 266 | 62 | ||
| AK027534.1 | - | 1 - 2 | 168 | 62 | ||
| BC013771.1 | - | 1 - 2 | 210 | 4385034 | 62 | |
| BI550730.1 | - | 1 - 2 | 227 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000884.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000359883.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000359883.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000359883.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000359883.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000359883.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000359883.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000359883.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000447596.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000447596.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000447596.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000447596.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000447596.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000447596.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09011797.hg.1 | TC09000884.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09011797.hg.1 | ENST00000359883.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09011797.hg.1 | ENST00000381809.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09011797.hg.1 | ENST00000474822.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09011797.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09011797.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09011797.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09011797.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012421.hg.1 | ENST00000539058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012421.hg.1 | ENST00000539058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007232.hg.1 | ENST00000432033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900164867.hg.1 | ENST00000359883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900164867.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900164867.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900164867.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900164867.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900164867.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900164867.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900164867.hg.1 | ENST00000359883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900164867.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900164867.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900164867.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900164867.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900164867.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900164867.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900164867.hg.1 | TC0900009437.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000007232.hg.1 | ENST00000432033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012421.hg.1 | ENST00000539058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012421.hg.1 | ENST00000539058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900164866.hg.1 | ENST00000359883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900164866.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900164866.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900164866.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900164866.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900164866.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900164866.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900164866.hg.1 | ENST00000359883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900164866.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900164866.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900164866.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900164866.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900164866.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900164866.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900164866.hg.1 | TC0900009437.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000359883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000359883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000359883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000359883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000359883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000359883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000359883.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000381809.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000447596.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000447596.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000447596.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000447596.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000447596.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000447596.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | ENST00000611749.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199852.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199853.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199855.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_001199856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_016282.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009437.hg.1 | NM_016282.3 | Refseq |

AK3
Genomic Map