Assay Details
| Gene Symbol: KPNA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002268.4 | NP_002259.1 | 1 - 2 | 372 | 150 | |
| GenBank mRNA | AB002533.2 | - | 1 - 2 | 75 | 150 | |
| AK291041.1 | - | 1 - 2 | 324 | 150 | ||
| BC028691.2 | - | 1 - 2 | 317 | 4838709 | 150 | |
| BC034493.2 | - | 1 - 2 | 372 | 4821602 | 150 | |
| U93240.1 | - | 1 - 2 | 216 | 150 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001956.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009883.hg.1 | ENST00000396110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009883.hg.1 | ENST00000396110.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035011.hg.1 | TC03001956.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035011.hg.1 | ENST00000334256.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035011.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035011.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035009.hg.1 | TC03001956.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035009.hg.1 | ENST00000334256.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035009.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008695.hg.1 | ENST00000402742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008695.hg.1 | ENST00000402742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008695.hg.1 | ENST00000402742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008695.hg.1 | ENST00000402742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008695.hg.1 | ENST00000402742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000483437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000483437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000483437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000483437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035012.hg.1 | TC03001956.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035012.hg.1 | ENST00000334256.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035012.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035012.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196980.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196980.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196980.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196980.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196980.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196980.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196980.hg.1 | TC0300012970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600008695.hg.1 | ENST00000402742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008695.hg.1 | ENST00000402742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008695.hg.1 | ENST00000402742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008695.hg.1 | ENST00000402742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008695.hg.1 | ENST00000402742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196978.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196978.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196978.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196978.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196978.hg.1 | TC0300012970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300196981.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196981.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196981.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196981.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196981.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196981.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196981.hg.1 | TC0300012970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300196982.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196982.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196982.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196982.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196982.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196982.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196982.hg.1 | TC0300012970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00009883.hg.1 | ENST00000396110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009883.hg.1 | ENST00000396110.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000483437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000483437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000483437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000483437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 111685 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 209079 | - | - |

KPNA4
Genomic Map