Assay Details
| Gene Symbol: KPNA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002268.4 | NP_002259.1 | 7 - 8 | 772 | 70 | |
| GenBank mRNA | AB002533.2 | - | 7 - 8 | 475 | 70 | |
| AK291041.1 | - | 7 - 8 | 724 | 70 | ||
| BC028691.2 | - | 7 - 8 | 717 | 4838709 | 70 | |
| BC034493.2 | - | 7 - 8 | 772 | 4821602 | 70 | |
| U93240.1 | - | 7 - 8 | 616 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001956.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000483437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000483437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000483437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | ENST00000483437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035002.hg.1 | TC03001956.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035002.hg.1 | ENST00000334256.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035002.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035003.hg.1 | TC03001956.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035003.hg.1 | ENST00000334256.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035003.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196972.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196972.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196972.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196972.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196972.hg.1 | TC0300012970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300196971.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196971.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196971.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196971.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196971.hg.1 | TC0300012970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000334256.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000469804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000483437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000483437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000483437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | ENST00000483437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012970.hg.1 | NM_002268.4 | Refseq |

KPNA4
Genomic Map