Assay Details
| Gene Symbol: HLTF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318934.1 | NP_001305863.1 | 12 - 13 | 1500 | 99 | |
| XM_011513093.1 | XP_011511395.1 | 12 - 13 | 1500 | 99 | ||
| XM_017007078.1 | XP_016862567.1 | 12 - 13 | 1479 | 99 | ||
| XM_017007079.1 | XP_016862568.1 | 12 - 13 | 1500 | 99 | ||
| GenBank mRNA | BC044659.1 | - | 12 - 13 | 1468 | 6015181 | 99 |
More Information
Set Membership: |
|

HLTF
Genomic Map