Assay Details
| Gene Symbol: IGF2-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028043.2 | 2 - 3 | 717 | 81 | ||
| NR_133657.1 | 2 - 3 | 606 | 81 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000058.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11001131.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11001131.hg.1 | TC11000058.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381363.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381363.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381363.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381363.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381363.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000381363.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11001130.hg.1 | ENST00000381363.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11001130.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11001130.hg.1 | TC11000058.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11001134.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11001134.hg.1 | TC11000058.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100145820.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145820.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145820.hg.1 | ENST00000381363.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145820.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145820.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145820.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145820.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145820.hg.1 | ENST00000381363.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145820.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145820.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145820.hg.1 | TC1100006561.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100145823.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145823.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145823.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145823.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145823.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145823.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145823.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145823.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145823.hg.1 | TC1100006561.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100145821.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145821.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145821.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145821.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145821.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145821.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145821.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145821.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145821.hg.1 | TC1100006561.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100145822.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145822.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145822.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145822.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145822.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145822.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145822.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145822.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145822.hg.1 | TC1100006561.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381363.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381363.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381363.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381363.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381363.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000381363.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006561.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145824.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145824.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145824.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145824.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145824.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145824.hg.1 | ENST00000381361.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145824.hg.1 | ENST00000445504.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100145824.hg.1 | NR_028043.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100145824.hg.1 | TC1100006561.hg.1 | Clariom D TC |

IGF2-AS
Genomic Map