Assay Details
| Gene Symbol: ENTPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248.3 | NP_001239.2 | 7 - 8 | 952 | 102 | |
| NM_001291960.1 | NP_001278889.1 | 7 - 8 | 993 | 102 | ||
| NM_001291961.1 | NP_001278890.1 | 7 - 8 | 952 | 102 | ||
| GenBank mRNA | AF034840.2 | - | 7 - 8 | 913 | 102 | |
| AF039917.1 | - | 7 - 8 | 916 | 102 | ||
| AK313322.1 | - | 7 - 8 | 978 | 102 | ||
| BC029869.1 | - | 6 - 7 | 856 | 5174918 | 102 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000208.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03004129.hg.1 | TC03000208.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004129.hg.1 | ENST00000301825.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004129.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004129.hg.1 | ENST00000456402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03004129.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300150008.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150008.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150008.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150008.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300150008.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300150008.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300150008.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150008.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150008.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300150008.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300150008.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300150008.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300150008.hg.1 | TC0300007117.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000301825.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000439533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000445129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | ENST00000456402.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001248.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291960.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007117.hg.1 | NM_001291961.1 | Refseq |

ENTPD3
Genomic Map