Assay Details
| Gene Symbol: FKBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002013.3 | NP_002004.1 | 1 - 2 | 179 | 107 | |
| XM_017021092.1 | XP_016876581.1 | 1 - 2 | 128 | 107 | ||
| GenBank mRNA | AK222993.1 | - | 1 - 2 | 128 | 107 | |
| AK311915.1 | - | 1 - 2 | 117 | 107 | ||
| BC016288.1 | - | 2 - 3 | 517 | 3919642 | 107 | |
| BC020809.1 | - | 1 - 2 | 117 | 4292678 | 107 | |
| BI598048.1 | - | 1 - 2 | 184 | 107 | ||
| BT006904.1 | - | 1 - 2 | 106 | 107 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001084.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000556231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000556231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000556231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000556231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017853.hg.1 | ENST00000423792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017853.hg.1 | ENST00000423792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012222.hg.1 | ENST00000216330.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012222.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012222.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012222.hg.1 | TC14001084.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012225.hg.1 | ENST00000216330.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012225.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012225.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012225.hg.1 | TC14001084.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013765.hg.1 | ENST00000424719.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012224.hg.1 | ENST00000216330.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012224.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012224.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012224.hg.1 | TC14001084.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012223.hg.1 | ENST00000216330.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012223.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012223.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012223.hg.1 | TC14001084.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400166617.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166617.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166617.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166617.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166617.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166617.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166617.hg.1 | TC1400009065.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000556231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000556231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000556231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000556231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009065.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166614.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166614.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166614.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166614.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166614.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166614.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166614.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166614.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166614.hg.1 | TC1400009065.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400166618.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166618.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166618.hg.1 | TC1400009065.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400166619.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166619.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166619.hg.1 | TC1400009065.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100013765.hg.1 | ENST00000424719.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017853.hg.1 | ENST00000423792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017853.hg.1 | ENST00000423792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166610.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166610.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166610.hg.1 | ENST00000556231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166610.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166610.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166610.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166610.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166610.hg.1 | ENST00000556231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166610.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166610.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166610.hg.1 | TC1400009065.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400166608.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166608.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166608.hg.1 | ENST00000556231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166608.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166608.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166608.hg.1 | ENST00000216330.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166608.hg.1 | ENST00000396062.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166608.hg.1 | ENST00000556231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166608.hg.1 | ENST00000557324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166608.hg.1 | NM_002013.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166608.hg.1 | TC1400009065.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 120058 | - | - |

FKBP3
Genomic Map