Assay Details
| Gene Symbol: EEA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003566.3 | NP_003557.2 | 10 - 11 | 1181 | 185 | |
| XM_011538814.2 | XP_011537116.1 | 11 - 12 | 1287 | 185 | ||
| XM_011538815.2 | XP_011537117.1 | 10 - 11 | 998 | 185 | ||
| XM_017020018.1 | XP_016875507.1 | 10 - 11 | 949 | 185 | ||
| GenBank mRNA | AK292008.1 | - | 10 - 11 | 1100 | 185 | |
| AK307918.1 | - | 10 - 11 | 1102 | 185 | ||
| X78998.1 | - | 10 - 11 | 987 | 185 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001811.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12023927.hg.1 | ENST00000322349.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12023927.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12023927.hg.1 | TC12001811.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200189307.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200189307.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200189307.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200189307.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200189307.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200189307.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200189307.hg.1 | TC1200011519.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |

EEA1
Genomic Map