Assay Details
| Gene Symbol: EEA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003566.3 | NP_003557.2 | 15 - 16 | 2190 | 133 | |
| XM_011538814.2 | XP_011537116.1 | 16 - 17 | 2296 | 133 | ||
| XM_011538815.2 | XP_011537117.1 | 15 - 16 | 2007 | 133 | ||
| XM_017020018.1 | XP_016875507.1 | 15 - 16 | 1958 | 133 | ||
| GenBank mRNA | AK292008.1 | - | 15 - 16 | 2109 | 133 | |
| AK307918.1 | - | 15 - 16 | 2111 | 133 | ||
| L40157.1 | - | 15 - 16 | 2061 | 133 | ||
| X78998.1 | - | 15 - 16 | 1996 | 133 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001811.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12023922.hg.1 | ENST00000322349.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12023922.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12023922.hg.1 | TC12001811.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200189301.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200189301.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200189301.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200189301.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200189301.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200189301.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200189301.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200189301.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200189301.hg.1 | TC1200011519.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |

EEA1
Genomic Map