Assay Details
| Gene Symbol: EEA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003566.3 | NP_003557.2 | 4 - 5 | 561 | 103 | |
| XM_011538814.2 | XP_011537116.1 | 5 - 6 | 667 | 103 | ||
| XM_011538815.2 | XP_011537117.1 | 4 - 5 | 378 | 103 | ||
| XM_017020018.1 | XP_016875507.1 | 4 - 5 | 329 | 103 | ||
| GenBank mRNA | AK292008.1 | - | 4 - 5 | 480 | 103 | |
| AK307918.1 | - | 4 - 5 | 482 | 103 | ||
| BC017837.1 | - | 4 - 5 | 461 | 103 | ||
| L40157.1 | - | 4 - 5 | 432 | 103 | ||
| X78998.1 | - | 4 - 5 | 367 | 103 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001811.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12023934.hg.1 | ENST00000322349.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12023934.hg.1 | ENST00000540777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12023934.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12023934.hg.1 | TC12001811.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000547833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000549790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | ENST00000553019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011519.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200189313.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200189313.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200189313.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200189313.hg.1 | ENST00000322349.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200189313.hg.1 | ENST00000418984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200189313.hg.1 | NM_003566.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200189313.hg.1 | TC1200011519.hg.1 | Clariom D TC |

EEA1
Genomic Map