Assay Details
| Gene Symbol: TRMT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031712.2 | NP_001026882.2 | 10 - 11 | 1131 | 121 | |
| XM_017011168.1 | XP_016866657.1 | 9 - 10 | 939 | 121 | ||
| XM_017011169.1 | XP_016866658.1 | 9 - 10 | 1101 | 121 | ||
| GenBank mRNA | AF182423.1 | - | 12 - 13 | 1262 | 121 | |
| AF532977.1 | - | 10 - 11 | 1096 | 121 | ||
| BC056878.1 | - | 10 - 11 | 1131 | 4443778 | 121 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000964.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000334379.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000368332.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000444121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000444121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000444121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000444121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000444121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000444121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000444121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000444121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000444121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000446681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000446681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000446681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000446681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000446681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000446681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000446681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000446681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000446681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000446681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000453993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000453993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000453993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000453993.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000461129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000461129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000461129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000461129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000461129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000461129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000461129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000461129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000461129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000461129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000461129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000466316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000466316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000466316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000466316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000466316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000466316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000466316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000468097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000468097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000468097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000468097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000468097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000468097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000468097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000468097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000468097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000468097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000468097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000468097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000468097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000468097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000473273.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000473273.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000473273.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000473273.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000479748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000489934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000489934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | ENST00000489934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009370.hg.1 | NM_001031712.2 | Refseq |

TRMT11
Genomic Map