Assay Details
| Gene Symbol: ABCD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005050.3 | NP_005041.1 | 3 - 4 | 436 | 61 | |
| XM_005267939.3 | XP_005267996.1 | 3 - 4 | 369 | 61 | ||
| XM_005267940.3 | XP_005267997.1 | 2 - 3 | 248 | 61 | ||
| XM_006720223.2 | XP_006720286.1 | 2 - 3 | 248 | 61 | ||
| XM_017021531.1 | XP_016877020.1 | 3 - 4 | 369 | 61 | ||
| XM_017021532.1 | XP_016877021.1 | 3 - 4 | 369 | 61 | ||
| XM_017021533.1 | XP_016877022.1 | 3 - 4 | 369 | 61 | ||
| XM_017021535.1 | XP_016877024.1 | 3 - 4 | 369 | 61 | ||
| XM_017021538.1 | XP_016877027.1 | 2 - 3 | 195 | 61 | ||
| XM_017021541.1 | XP_016877030.1 | 3 - 4 | 306 | 61 | ||
| XM_017021544.1 | XP_016877033.1 | 3 - 4 | 369 | 61 | ||
| GenBank mRNA | AF009746.1 | - | 3 - 4 | 335 | 61 | |
| AK291332.1 | - | 3 - 4 | 306 | 61 | ||
| AK297934.1 | - | 2 - 3 | 222 | 61 | ||
| BC012815.2 | - | 3 - 4 | 290 | 2960427 | 61 | |
| BI916327.1 | - | 3 - 4 | 428 | 61 | ||
| BT007412.1 | - | 3 - 4 | 284 | 61 | ||
| CR457104.1 | - | 3 - 4 | 284 | 61 | ||
| DC332064.1 | - | 3 - 4 | 436 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK297934.1, XM_005267939.3, XM_005267940.3, XM_006720223.2, XM_017021535.1, XM_017021538.1, XM_017021541.1, XM_017021544.1 transcripts.
Set Membership: |
|

ABCD4
Genomic Map