Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014913.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014913.hg.1 |
ENST00000374633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014913.hg.1 |
ENST00000467975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014913.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014913.hg.1 |
TC10001188.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014915.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014915.hg.1 |
ENST00000374633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014915.hg.1 |
ENST00000467975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014915.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014915.hg.1 |
TC10001188.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000467975.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000467975.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000467975.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000467975.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000467975.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000467975.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
ENST00000467975.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010367.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000467975.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000467975.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000467975.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000467975.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000467975.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000467975.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
ENST00000467975.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010367.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000170511.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170511.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170511.hg.1 |
ENST00000467975.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170511.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000170511.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170511.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170511.hg.1 |
ENST00000467975.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170511.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000170511.hg.1 |
TC1000010367.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000170514.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170514.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170514.hg.1 |
ENST00000467975.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170514.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000170514.hg.1 |
ENST00000302609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170514.hg.1 |
ENST00000374633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170514.hg.1 |
ENST00000467975.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170514.hg.1 |
NM_145011.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000170514.hg.1 |
TC1000010367.hg.1 |
Clariom D TC |