Assay Details
| Gene Symbol: MYLK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308301.1 | NP_001295230.1 | 10 - 11 | 1567 | 63 | |
| NM_182493.2 | NP_872299.2 | 11 - 12 | 2386 | 63 | ||
| XM_006721334.3 | XP_006721397.1 | 11 - 12 | 1854 | 63 | ||
| GenBank mRNA | AB451455.1 | - | 11 - 12 | 2270 | 63 | |
| AJ247087.1 | - | 11 - 12 | 2386 | 63 | ||
| AK299443.1 | - | 10 - 11 | 1567 | 63 | ||
| BC109097.1 | - | 11 - 12 | 2283 | 40034724 | 63 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001085.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of vascular permeability involved in acute inflammatory response
protein phosphorylation
sarcomere organization
sarcomerogenesis
cardiac myofibril assembly
positive regulation of sarcomere organization
cellular response to interleukin-1
calmodulin-dependent protein kinase activity
myosin light chain kinase activity
ATP binding
regulation of vascular permeability involved in acute inflammatory response
protein phosphorylation
sarcomere organization
sarcomerogenesis
cardiac myofibril assembly
positive regulation of sarcomere organization
cellular response to interleukin-1
calmodulin-dependent protein kinase activity
myosin light chain kinase activity
ATP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000562104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000562104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000562104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000562104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000562218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000564731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000564731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000564731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000564731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000564731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000564731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000564731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000564731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000565182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000565182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000565182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000565182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000569810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000569810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000569810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000569844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000569844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000569844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | ENST00000569844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015240.hg.1 | ENST00000394809.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015240.hg.1 | ENST00000536476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015240.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015240.hg.1 | TC16001085.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000562104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000562104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000562104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000562104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000562218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000564731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000564731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000564731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000564731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000564731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000564731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000564731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000564731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000565182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000565182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000565182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000565182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000569810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000569810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000569810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000569844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000569844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000569844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | ENST00000569844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010174.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178396.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178396.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178396.hg.1 | ENST00000562104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178396.hg.1 | ENST00000565182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178396.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178396.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178396.hg.1 | ENST00000394809.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178396.hg.1 | ENST00000536476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178396.hg.1 | ENST00000562104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178396.hg.1 | ENST00000565182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178396.hg.1 | NM_001308301.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178396.hg.1 | NM_182493.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178396.hg.1 | TC1600010174.hg.1 | Clariom D TC |

MYLK3
Genomic Map