Assay Details
| Gene Symbol: ERP27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152321.3 | NP_689534.1 | 1 - 2 | 129 | 78 | |
| GenBank mRNA | AK056677.1 | - | 1 - 2 | 129 | 78 | |
| AY358536.1 | - | 1 - 2 | 109 | 78 | ||
| BC030218.1 | - | 1 - 2 | 101 | 5207225 | 78 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001277.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12016638.hg.1 | ENST00000266397.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016638.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016638.hg.1 | TC12001277.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000540097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000540097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000540097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000540097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000540097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000540097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000540097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000540097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000544881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000544881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | ENST00000544881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_001300784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_001300784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_001300784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_001300784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_001300784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_001300784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_001300784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_001300784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_001300784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016637.hg.1 | ENST00000266397.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016637.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016637.hg.1 | TC12001277.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200175719.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175719.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175719.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175719.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175719.hg.1 | TC1200010023.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200175721.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175721.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175721.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175721.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175721.hg.1 | TC1200010023.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000540097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000540097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000540097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000540097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000540097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000540097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000540097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000540097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000544881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000544881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | ENST00000544881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_001300784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_001300784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_001300784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_001300784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_001300784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_001300784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_001300784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_001300784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_001300784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010023.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175718.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175718.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175718.hg.1 | ENST00000266397.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175718.hg.1 | NM_152321.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175718.hg.1 | TC1200010023.hg.1 | Clariom D TC |

ERP27
Genomic Map