| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
ENST00000531884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
ENST00000531884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
ENST00000531884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
ENST00000531884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12008257.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12008257.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12008257.hg.1 |
TC12000621.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12008256.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12008256.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12008256.hg.1 |
TC12000621.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12008259.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12008259.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12008259.hg.1 |
TC12000621.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12008258.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12008258.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12008258.hg.1 |
TC12000621.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1200159972.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200159972.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200159972.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200159972.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200159972.hg.1 |
TC1200008165.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200159971.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200159971.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200159971.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200159971.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200159971.hg.1 |
TC1200008165.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
ENST00000531884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
ENST00000531884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
ENST00000531884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
ENST00000531884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008165.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200159973.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200159973.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200159973.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200159973.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200159973.hg.1 |
TC1200008165.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200159974.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200159974.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200159974.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200159974.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200159974.hg.1 |
TC1200008165.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200159976.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200159976.hg.1 |
ENST00000531884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200159976.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200159976.hg.1 |
ENST00000258111.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200159976.hg.1 |
ENST00000531884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200159976.hg.1 |
NM_014505.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200159976.hg.1 |
TC1200008165.hg.1 |
Clariom D TC |