Assay Details
| Gene Symbol: BHMG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310124.1 | NP_001297053.1 | 1 - 2 | 414 | 54 | |
| GenBank mRNA | BI463439.1 | - | 1 - 2 | 425 | 54 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001310124.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164055.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164055.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164055.hg.1 | TC1900008345.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900164055.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164055.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164054.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164054.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164054.hg.1 | TC1900008345.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900164054.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164054.hg.1 | NM_001310124.1 | Refseq |

BHMG1
Genomic Map