Assay Details
| Gene Symbol: BHMG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310124.1 | NP_001297053.1 | 8 - 9 | 74 | ||
| XM_011527571.2 | XP_011525873.1 | 8 - 9 | 74 | |||
| XM_011527573.2 | XP_011525875.1 | 7 - 8 | 74 | |||
| XM_011527574.2 | XP_011525876.1 | 7 - 8 | 74 | |||
| GenBank mRNA | DV080301.1 | - | 4 - 5 | 74 | ||
| DV080302.1 | - | 4 - 5 | 389 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008345.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164065.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164065.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164065.hg.1 | TC1900008345.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900164065.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164065.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164066.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164066.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164066.hg.1 | TC1900008345.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900164066.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164066.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164063.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164063.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164063.hg.1 | TC1900008345.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900164063.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164063.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164062.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164062.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164062.hg.1 | TC1900008345.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900164062.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164062.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164064.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164064.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164064.hg.1 | TC1900008345.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900164064.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164064.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164067.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164067.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164067.hg.1 | TC1900008345.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900164067.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164067.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164068.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164068.hg.1 | NM_001310124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900164068.hg.1 | TC1900008345.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900164068.hg.1 | ENST00000457052.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900164068.hg.1 | NM_001310124.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 244123_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 244123_at | - | - |
| Human Genome U133 Plus 2.0 Array | 241822_at | - | - |
| Human Genome U133 Plus 2.0 Array | 241822_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241822_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244123_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241822_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241822_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241822_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244123_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241822_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244123_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241822_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241822_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241822_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244123_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244123_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241822_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244123_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244123_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241822_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241822_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244123_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244123_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244123_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 244123_at2 | - | - |
| Human Genome U95 Set | 81033_at | - | - |
| Human Genome U95 Set | 81033_PM_at | - | - |
| Human Genome U95 Set | 75193_f_at | - | - |
| Human Genome U95 Set | 75191_PM_i_at | - | - |
| Human Genome U95 Set | 75191_i_at | - | - |
| Human Genome U95 Set | 75193_PM_f_at | - | - |
| Human Genome U133 Set | 244123_PM_at | - | - |
| Human Genome U133 Set | 241822_at | - | - |
| Human Genome U133 Set | 244123_at | - | - |
| Human Genome U133 Set | 241822_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3217473 | - | - |
| HumanHT-12 v4 Expression BeadChip | ILMN_3282917 | - | - |

BHMG1
Genomic Map