Assay Details
| Gene Symbol: MLKL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152649.3 | NP_689862.1 | 2 - 3 | 1148 | 135 | |
| XM_005255834.3 | XP_005255891.1 | 3 - 4 | 1047 | 135 | ||
| XM_011522936.2 | XP_011521238.1 | 2 - 3 | 1187 | 135 | ||
| GenBank mRNA | AK091708.1 | - | 2 - 3 | 891 | 135 | |
More Information
Set Membership: |
|

MLKL
Genomic Map