Assay Details
| Gene Symbol: ARRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017017753.1 | XP_016873242.1 | 1 - 2 | 146 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017017753.1 transcripts.
Set Membership: |
|

ARRB1
Genomic Map