Assay Details
Gene Symbol: MARVELD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038603.2 | NP_001033692.2 | 3 - 4 | 1256 | 144 | |
XM_005248445.4 | XP_005248502.1 | 3 - 4 | 1550 | 144 | ||
XM_005248446.4 | XP_005248503.1 | 3 - 4 | 1344 | 144 | ||
GenBank mRNA | AB219936.1 | - | 2 - 3 | 1182 | 144 | |
AB219937.1 | - | 2 - 3 | 1182 | 144 | ||
AK055094.1 | - | 3 - 4 | 1252 | 144 | ||
AK292962.1 | - | 3 - 4 | 1240 | 144 | ||
BC033689.1 | - | 3 - 4 | 1241 | 5210373 | 144 | |
DQ682656.1 | - | 3 - 4 | 1241 | 144 | ||
DQ682658.1 | - | 3 - 4 | 852 | 144 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05000303.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000515844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | ENST00000515844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05004059.hg.1 | TC05000303.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05004059.hg.1 | ENST00000282886.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05004059.hg.1 | ENST00000325631.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05004059.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05004059.hg.1 | ENST00000436532.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05004059.hg.1 | ENST00000454295.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05004059.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05004059.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152068.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152068.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152068.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152068.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152068.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152068.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152068.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152068.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152068.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152068.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152068.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152068.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500152068.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152068.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500152068.hg.1 | TC0500007676.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000325631.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000413223.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000436532.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000454295.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000512803.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000515844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | ENST00000515844.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001038603.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500007676.hg.1 | NM_001244734.1 | Refseq |
