Assay Details
| Gene Symbol: HAND2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BF313018.1 | - | 2 - 3 | 544 | 150 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000503024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000503024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000503024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000503024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000503024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000503024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000505300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000505300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000505300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000505300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000505300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000505300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000505300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000505300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000621866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000621866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000621866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000621866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000621866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000621866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | ENST00000621866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000359562.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000503024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000503024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000503024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000503024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000503024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000503024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000505300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000505300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000505300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000505300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000505300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000505300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000505300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000505300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000621866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000621866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000621866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000621866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000621866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000621866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | ENST00000621866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | NM_021973.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012443.hg.1 | NM_021973.2 | Refseq |

HAND2
Genomic Map