| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
ENST00000374919.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
ENST00000374919.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
ENST00000374919.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
ENST00000374919.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
ENST00000374919.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
ENST00000475785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
ENST00000475785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
ENST00000475785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
ENST00000475785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
ENST00000475785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
ENST00000475785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
ENST00000475785.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
ENST00000478405.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
ENST00000478405.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
ENST00000478405.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
ENST00000478405.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
ENST00000478405.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
ENST00000478405.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
NM_001010862.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
NM_001010862.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
NM_001010862.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
NM_001010862.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
NM_001010862.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
NM_001010862.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
NM_001010862.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
NR_027139.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
NR_027139.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
NR_027139.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
NR_027139.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
NR_027139.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
NR_027139.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009836.hg.1 |
NR_027139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
ENST00000374919.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
ENST00000374919.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
ENST00000374919.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
ENST00000374919.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
ENST00000374919.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
ENST00000475785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
ENST00000475785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
ENST00000475785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
ENST00000475785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
ENST00000475785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
ENST00000475785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
ENST00000475785.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
ENST00000478405.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
ENST00000478405.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
ENST00000478405.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
ENST00000478405.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
ENST00000478405.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
ENST00000478405.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
NM_001010862.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
NM_001010862.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
NM_001010862.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
NM_001010862.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
NM_001010862.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
NM_001010862.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
NM_001010862.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
NR_027139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
NR_027139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
NR_027139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
NR_027139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
NR_027139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
NR_027139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009836.hg.1 |
NR_027139.1 |
Refseq |