Assay Details
| Gene Symbol: NDST4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022569.2 | NP_072091.1 | 8 - 9 | 2497 | 115 | |
| XM_017008545.1 | XP_016864034.1 | 7 - 8 | 1495 | 115 | ||
| XM_017008546.1 | XP_016864035.1 | 6 - 7 | 801 | 115 | ||
| GenBank mRNA | AB036429.1 | - | 8 - 9 | 2497 | 115 | |
| AK289670.1 | - | 8 - 9 | 2475 | 115 | ||
| BC113075.2 | - | 6 - 7 | 1086 | 115 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001488.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04021119.hg.1 | TC04001488.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021119.hg.1 | ENST00000264363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021119.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400180073.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400180073.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400180073.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400180073.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400180073.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400180073.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400180073.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400180073.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400180073.hg.1 | TC0400011659.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000264363.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000504854.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000514570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | ENST00000613194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011659.hg.1 | NM_022569.2 | Refseq |

NDST4
Genomic Map