Assay Details
| Gene Symbol: CDC25C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287582.1 | NP_001274511.1 | 6 - 7 | 778 | 149 | |
| NM_001287583.1 | NP_001274512.1 | 6 - 7 | 895 | 149 | ||
| NM_001318098.1 | NP_001305027.1 | 6 - 7 | 899 | 149 | ||
| NM_001790.4 | NP_001781.2 | 6 - 7 | 782 | 149 | ||
| XM_005272145.3 | XP_005272202.1 | 6 - 7 | 895 | 149 | ||
| XM_006714739.3 | XP_006714802.1 | 5 - 6 | 493 | 149 | ||
| XM_011543759.2 | XP_011542061.1 | 7 - 8 | 566 | 149 | ||
| XM_011543760.2 | XP_011542062.1 | 6 - 7 | 895 | 149 | ||
| XM_011543763.1 | XP_011542065.1 | 4 - 5 | 191 | 149 | ||
| XM_017010101.1 | XP_016865590.1 | 6 - 7 | 773 | 149 | ||
| GenBank mRNA | AF277725.1 | - | 3 - 4 | 239 | 149 | |
| AK097710.1 | - | 6 - 7 | 895 | 149 | ||
| AK301828.1 | - | 7 - 8 | 845 | 149 | ||
| BC019089.2 | - | 6 - 7 | 672 | 4858949 | 149 | |
| M34065.1 | - | 6 - 7 | 671 | 149 | ||
More Information
Set Membership: |
|

CDC25C
Genomic Map