Assay Details
Gene Symbol: QSER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076786.2 | NP_001070254.1 | 6 - 7 | 4560 | 195 | |
XM_017018327.1 | XP_016873816.1 | 6 - 7 | 4912 | 195 | ||
XM_017018328.1 | XP_016873817.1 | 6 - 7 | 4912 | 195 | ||
XM_017018329.1 | XP_016873818.1 | 4 - 5 | 5365 | 195 | ||
XM_017018330.1 | XP_016873819.1 | 5 - 6 | 4310 | 195 | ||
GenBank mRNA | AK025577.1 | - | 3 - 4 | 1287 | 195 | |
AK123858.1 | - | 3 - 4 | 2622 | 195 | ||
AK125902.1 | - | 3 - 4 | 2108 | 195 | ||
AK126023.1 | - | 7 - 8 | 3864 | 195 | ||
AL834141.1 | - | 3 - 4 | 3256 | 195 | ||
BC112935.1 | - | 6 - 7 | 4560 | 195 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527250.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527250.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527250.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527250.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000528034.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000528034.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000528155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000528155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000528155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000528155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | ENST00000528155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100150801.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100150801.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100150801.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100150801.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100150801.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100150801.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100150801.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100150801.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100150801.hg.1 | TC1100007217.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000399302.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000524678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527250.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527250.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527250.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527250.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000527788.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000528034.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000528034.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000528155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000528155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000528155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000528155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | ENST00000528155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007217.hg.1 | NM_001076786.2 | Refseq |
