Assay Details
| Gene Symbol: HEXA-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027262.1 | 1 | 1747 | 86 | ||
| GenBank mRNA | AK023377.1 | - | 1 | 1495 | 86 | |
| BC068211.1 | - | 1 | 1747 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | hexosaminidase subunit alpha | HEXA | TSD | -187.0 | Anti-sense | Chr. 15 - 72343435 - 72376473 on NCBI Build GRCh38 |
| Downstream | transmembrane protein 202 | TMEM202 | 20.2kb | Sense | Chr. 15 - 72398320 - 72408367 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09751537 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30244843 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09751541 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30244839 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09751532 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30244840 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30244845 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30244834 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30244838 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09751534 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09751543 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09751546 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 49284 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 11485 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 59979 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 370271 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3315149 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 43087 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 2987 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3315149 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3315149 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751458 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244771 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244766 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16803232 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751512 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751513 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751478 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16803218 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751464 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244775 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244778 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244821 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751491 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244779 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751477 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244741 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751446 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751454 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244822 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244737 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244763 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244820 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244748 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751516 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751494 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244754 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244772 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244759 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244736 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244761 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244813 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751461 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244807 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751449 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751522 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244774 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244801 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751490 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244768 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751471 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244745 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751524 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751444 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751525 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751463 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751514 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751450 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751486 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244744 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244774 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244775 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751524 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244737 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751525 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751516 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751512 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244768 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751494 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751477 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244801 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244807 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751513 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244761 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751478 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244744 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16803232 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244820 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751458 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244822 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751522 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244779 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244748 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751454 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751514 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244736 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244759 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244771 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244772 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16803218 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751490 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751449 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751444 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751491 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244741 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244763 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751450 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244778 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751486 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244766 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244813 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751463 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244754 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751461 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751471 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751446 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244745 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30244821 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09751464 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P37455 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P117425 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P37455 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 14965 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 53257 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P117425 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751480 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751457 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751491 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751545 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751530 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751453 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751443 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751445 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751449 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751519 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751486 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751490 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751515 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751514 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751523 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751468 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751526 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751509 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751447 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751543 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751541 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751471 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16803215 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16803232 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751538 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16803221 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751547 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751537 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16803218 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751484 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751528 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751539 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751524 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751513 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751462 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751454 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16803214 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751488 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751477 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751511 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751474 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751522 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751549 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751476 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751516 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751466 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751460 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09751533 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P117425 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7984634 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7984633 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7984632 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7984631 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 220718_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007787.hg.1 | ENST00000567598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007787.hg.1 | ENST00000567598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007787.hg.1 | NR_027262.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007787.hg.1 | NR_027262.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500155245.hg.1 | ENST00000567598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500155245.hg.1 | NR_027262.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500155245.hg.1 | ENST00000567598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500155245.hg.1 | NR_027262.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500155245.hg.1 | TC1500007787.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500007787.hg.1 | ENST00000567598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007787.hg.1 | ENST00000567598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007787.hg.1 | NR_027262.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007787.hg.1 | NR_027262.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 220718_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 220718_at | - | - |
| Human Genome U133A 2.0 Array | 220718_at | - | - |
| Human Genome U133A 2.0 Array | 220718_PM_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw595714 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220718_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220718_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220718_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220718_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220718_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220718_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220718_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220718_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220718_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220718_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220718_at8 | - | - |
| Human Genome U133 Set | 220718_PM_at | - | - |
| Human Genome U133 Set | 220718_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053041 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053041 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 198017 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3268535 | BC068211.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3268535 | NR_027262.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3268535 | AK023377.1 | human_transcriptome |

HEXA-AS1
Genomic Map