Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004090.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004090.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004090.hg.1 |
TC10000350.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004092.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004092.hg.1 |
ENST00000467359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004092.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004092.hg.1 |
TC10000350.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000467359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000467359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000467359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000467359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000476752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000476752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000476752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000476752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000476752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000494277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000494277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000494277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
ENST00000494277.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152008.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152008.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152008.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152008.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152008.hg.1 |
TC1000007641.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000152011.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152011.hg.1 |
ENST00000467359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152011.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152011.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152011.hg.1 |
ENST00000467359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152011.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152011.hg.1 |
TC1000007641.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000467359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000467359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000467359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000467359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000476752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000476752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000476752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000476752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000476752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000494277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000494277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000494277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
ENST00000494277.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007641.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152009.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152009.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152009.hg.1 |
ENST00000373970.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152009.hg.1 |
NM_012242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152009.hg.1 |
TC1000007641.hg.1 |
Clariom D TC |