Assay Details
| Gene Symbol: NDUFB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004545.3 | NP_004536.2 | 1 - 2 | 164 | 84 | |
| GenBank mRNA | BC104672.1 | - | 1 - 2 | 84 | 84 | |
| BC126232.1 | - | 1 - 2 | 161 | 8991948 | 84 | |
| BC126234.1 | - | 1 - 2 | 161 | 8991950 | 84 | |
| BM052981.1 | - | 1 - 2 | 164 | 84 | ||
| BX476462.1 | - | 1 - 2 | 63 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001447.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300007321.hg.1 | ENST00000435191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016269.hg.1 | ENST00000329559.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016269.hg.1 | ENST00000553377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016269.hg.1 | ENST00000555441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016269.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016269.hg.1 | TC14001447.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016277.hg.1 | ENST00000329559.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016277.hg.1 | ENST00000556555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016277.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016277.hg.1 | TC14001447.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000556555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000556555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000556555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000556555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016278.hg.1 | ENST00000329559.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016278.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14016278.hg.1 | TC14001447.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400173955.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173955.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173955.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400173955.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173955.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173955.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400173955.hg.1 | TC1400009998.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400173943.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173943.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173943.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173943.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173943.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173943.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173943.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400173943.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173943.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173943.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173943.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173943.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173943.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173943.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400173943.hg.1 | TC1400009998.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400173951.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173951.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173951.hg.1 | ENST00000556555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173951.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173951.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173951.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400173951.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173951.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173951.hg.1 | ENST00000556555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173951.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173951.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173951.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400173951.hg.1 | TC1400009998.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000553666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000555441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000556555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000556555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000556555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000556555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009998.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400173954.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173954.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173954.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400173954.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173954.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173954.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400173954.hg.1 | TC1400009998.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400173952.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173952.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173952.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173952.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173952.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400173952.hg.1 | ENST00000329559.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173952.hg.1 | ENST00000553514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173952.hg.1 | ENST00000605997.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173952.hg.1 | ENST00000617122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400173952.hg.1 | NM_004545.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400173952.hg.1 | TC1400009998.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300007321.hg.1 | ENST00000435191.1 | Ensembl |

NDUFB1
Genomic Map