Assay Details
| Gene Symbol: SMAD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005905.5 | NP_005896.1 | 3 - 4 | 1011 | 136 | |
| XM_005266401.3 | XP_005266458.1 | 2 - 3 | 720 | 136 | ||
| GenBank mRNA | BC011559.1 | - | 2 - 3 | 808 | 4558912 | 136 |
| D83761.1 | - | 2 - 3 | 723 | 136 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ureteric bud development
transcription, DNA-templated
protein phosphorylation
transforming growth factor beta receptor signaling pathway
BMP signaling pathway
midbrain development
hindbrain development
cartilage development
bone development
SMAD protein signal transduction
cellular response to organic cyclic compound
cellular response to BMP stimulus
positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transforming growth factor beta receptor, pathway-specific cytoplasmic mediator activity
metal ion binding
ureteric bud development
transcription, DNA-templated
protein phosphorylation
transforming growth factor beta receptor signaling pathway
BMP signaling pathway
midbrain development
hindbrain development
cartilage development
bone development
SMAD protein signal transduction
cellular response to organic cyclic compound
cellular response to BMP stimulus
positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transforming growth factor beta receptor, pathway-specific cytoplasmic mediator activity
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000483941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000483941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000483941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000483941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | ENST00000483941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000350148.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000379826.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000399275.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000483941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000483941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000483941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000483941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | ENST00000483941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_001127217.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_005905.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008626.hg.1 | NM_005905.5 | Refseq |

SMAD9
Genomic Map