Assay Details
| Gene Symbol: ARHGEF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015320.3 | NP_056135.2 | 4 - 5 | 943 | 75 | |
| NM_032995.2 | NP_127462.1 | 3 - 4 | 532 | 75 | ||
| XM_005263681.1 | XP_005263738.1 | 4 - 5 | 4020 | 75 | ||
| XM_011511274.2 | XP_011509576.1 | 4 - 5 | 4020 | 75 | ||
| XM_017004231.1 | XP_016859720.1 | 4 - 5 | 4020 | 75 | ||
| GenBank mRNA | AB029035.1 | - | 3 - 4 | 498 | 75 | |
| AF249745.1 | - | 4 - 5 | 944 | 75 | ||
| DQ424905.1 | - | 2 - 3 | 424 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000828.hg.1
Set Membership: |
|

ARHGEF4
Genomic Map