Assay Details
| Gene Symbol: BLNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114094.1 | NP_001107566.1 | 7 - 8 | 786 | 78 | |
| NM_001258441.1 | NP_001245370.1 | 7 - 8 | 726 | 78 | ||
| NR_047681.1 | 6 - 7 | 567 | 78 | |||
| NR_047682.1 | 6 - 7 | 562 | 78 | |||
| XM_017016159.1 | XP_016871648.1 | 7 - 8 | 664 | 78 | ||
| GenBank mRNA | AF068181.1 | - | 7 - 8 | 761 | 78 | |
| AK225546.1 | - | 7 - 8 | 769 | 78 | ||
| AM180332.1 | - | 7 - 8 | 608 | 78 | ||
| AM180333.1 | - | 7 - 8 | 608 | 78 | ||
| AM180334.1 | - | 7 - 8 | 608 | 78 | ||
| AM180335.1 | - | 4 - 5 | 246 | 78 | ||
| AM180338.1 | - | 6 - 7 | 449 | 78 | ||
| AM180339.1 | - | 6 - 7 | 444 | 78 | ||
| AM180344.1 | - | 5 - 6 | 517 | 78 | ||
| AM180346.1 | - | 5 - 6 | 544 | 78 | ||
More Information
Set Membership: |
|

BLNK
Genomic Map