Assay Details
| Gene Symbol: IKBKAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011519136.1 | XP_011517438.1 | 31 - 32 | 3708 | 103 | |
| XM_011519137.1 | XP_011517439.1 | 31 - 32 | 3482 | 103 | ||
More Information
Set Membership: |
|

IKBKAP
Genomic Map