Assay Details
| Gene Symbol: IL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172348.2 | NP_758858.1 | 1 - 2 | 193 | 120 | |
| GenBank mRNA | AB102862.1 | - | 1 - 2 | 128 | 120 | |
| X81851.1 | - | 1 - 2 | 128 | 120 | ||
More Information
Set Membership: |
|

IL4
Genomic Map