Assay Details
| Gene Symbol: MCF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099855.1 | NP_001093325.1 | 4 - 5 | 393 | 114 | |
| NM_001171876.1 | NP_001165347.1 | 4 - 5 | 393 | 114 | ||
| XM_005262413.4 | XP_005262470.1 | 5 - 6 | 541 | 114 | ||
| XM_011531339.2 | XP_011529641.1 | 5 - 6 | 600 | 114 | ||
| XM_017029529.1 | XP_016885018.1 | 4 - 5 | 362 | 114 | ||
| XM_017029530.1 | XP_016885019.1 | 4 - 5 | 362 | 114 | ||
| XM_017029531.1 | XP_016885020.1 | 4 - 5 | 295 | 114 | ||
| GenBank mRNA | AK296529.1 | - | 4 - 5 | 493 | 114 | |
| AK302957.1 | - | 4 - 5 | 341 | 114 | ||
| AK316416.1 | - | 4 - 5 | 381 | 114 | ||
| AL117234.1 | - | 3 - 4 | 276 | 114 | ||
| DC298193.1 | - | 4 - 5 | 393 | 114 | ||
More Information
Set Membership: |
|

MCF2
Genomic Map