Assay Details
| Gene Symbol: HIRIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003609.4 | NP_003600.2 | 4 - 5 | 1734 | 80 | |
| GenBank mRNA | AJ223350.1 | - | 3 - 4 | 1112 | 80 | |
| AJ223351.2 | - | 4 - 5 | 1631 | 80 | ||
| AK312504.1 | - | 4 - 5 | 1250 | 80 | ||
| BC000588.2 | - | 4 - 5 | 1704 | 3345739 | 80 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001014.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16014517.hg.1 | ENST00000279392.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014517.hg.1 | ENST00000352552.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014517.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014517.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014517.hg.1 | TC16001014.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000565996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000565996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000565996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000565996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000566471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000566471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000566471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000566471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000568880.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000568880.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000568880.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000568880.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | ENST00000568880.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600176576.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600176576.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600176576.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600176576.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600176576.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600176576.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600176576.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600176576.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600176576.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600176576.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600176576.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600176576.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600176576.hg.1 | TC1600009947.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000279392.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000563680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000564026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000565996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000565996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000565996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000565996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000566471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000566471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000566471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000566471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000568880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000568880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000568880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000568880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | ENST00000568880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_001197323.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_003609.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009947.hg.1 | NM_003609.4 | Refseq |

HIRIP3
Genomic Map