Assay Details
| Gene Symbol: DHRS7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001220493.1 | NP_001207422.1 | 2 - 3 | 280 | 103 | |
| GenBank mRNA | BC171909.1 | - | 2 - 3 | 280 | 9054563 | 103 |
More Information
Set Membership: |
|

DHRS7C
Genomic Map