Assay Details
| Gene Symbol: MYH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002472.2 | NP_002463.2 | 28 - 29 | 3957 | 100 | |
| GenBank mRNA | M36769.1 | - | 27 - 28 | 3935 | 100 | |
| Z38133.1 | - | 27 - 28 | 3935 | 100 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001131.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
protein dephosphorylation
muscle contraction
muscle filament sliding
ATP metabolic process
microfilament motor activity
calmodulin binding
ATP binding
structural constituent of muscle
ATPase activity
myosin phosphatase activity
myosin light chain binding
actin filament binding
skeletal muscle contraction
protein dephosphorylation
muscle contraction
muscle filament sliding
ATP metabolic process
microfilament motor activity
calmodulin binding
ATP binding
structural constituent of muscle
ATPase activity
myosin phosphatase activity
myosin light chain binding
actin filament binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17015292.hg.1 | ENST00000252173.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015292.hg.1 | ENST00000403437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015292.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015292.hg.1 | TC17001131.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015293.hg.1 | ENST00000252173.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015293.hg.1 | ENST00000403437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015293.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015293.hg.1 | TC17001131.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015291.hg.1 | ENST00000252173.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015291.hg.1 | ENST00000403437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015291.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015291.hg.1 | TC17001131.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009742.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700176062.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700176062.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700176062.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700176062.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700176062.hg.1 | TC1700009742.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700176060.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700176060.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700176060.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700176060.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700176060.hg.1 | TC1700009742.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700176061.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700176061.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700176061.hg.1 | ENST00000403437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700176061.hg.1 | NM_002472.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700176061.hg.1 | TC1700009742.hg.1 | Clariom D TC |

MYH8
Genomic Map