Assay Details
| Gene Symbol: CASK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126054.2 | NP_001119526.1 | 19 - 20 | 1811 | 120 | |
| NM_001126055.2 | NP_001119527.1 | 18 - 19 | 1793 | 120 | ||
| XM_011543995.2 | XP_011542297.1 | 18 - 19 | 1863 | 120 | ||
| GenBank mRNA | AB039327.2 | - | 7 - 8 | 969 | 120 | |
| AF035582.1 | - | 18 - 19 | 1762 | 120 | ||
| AF262405.1 | - | 17 - 18 | 1669 | 120 | ||
| AK313399.1 | - | 18 - 19 | 1946 | 120 | ||
| BC117311.1 | - | 19 - 20 | 1784 | 40125862 | 120 | |
| BC143454.1 | - | 19 - 20 | 1784 | 9051962 | 120 | |
| BC143456.1 | - | 18 - 19 | 1766 | 9051964 | 120 | |
More Information
Set Membership: |
|

CASK
Genomic Map