Assay Details
| Gene Symbol: PBXIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020524.3 | NP_065385.2 | 1 - 2 | 95 | 78 | |
| GenBank mRNA | AF221521.1 | - | 1 - 2 | 44 | 78 | |
| AK300544.1 | - | 1 - 2 | 59 | 78 | ||
| AK303863.1 | - | 1 - 2 | 39 | 78 | ||
| AK310712.1 | - | 1 - 2 | 70 | 78 | ||
| AK315551.1 | - | 1 - 2 | 76 | 78 | ||
| HY108652.1 | - | 1 - 2 | 95 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003298.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF221521.1,AK300544.1,AK303863.1,NM_020524.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050922.hg.1 | TC01003298.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050922.hg.1 | ENST00000351146.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050922.hg.1 | ENST00000368460.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050922.hg.1 | ENST00000368463.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050922.hg.1 | ENST00000368465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050922.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050926.hg.1 | TC01003298.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050926.hg.1 | ENST00000351146.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050926.hg.1 | ENST00000368460.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050926.hg.1 | ENST00000368463.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050926.hg.1 | ENST00000498553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050926.hg.1 | ENST00000539880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050926.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219979.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219979.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219979.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219979.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219979.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219979.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219979.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219979.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219979.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219979.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219979.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219979.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219979.hg.1 | TC0100015932.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100219976.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219976.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219976.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219976.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219976.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219976.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219976.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219976.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219976.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219976.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219976.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219976.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219976.hg.1 | TC0100015932.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100219980.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219980.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219980.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219980.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219980.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219980.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219980.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219980.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219980.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219980.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219980.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219980.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219980.hg.1 | TC0100015932.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |

PBXIP1
Genomic Map