Assay Details
| Gene Symbol: PBXIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020524.3 | NP_065385.2 | 2 - 3 | 189 | 60 | |
| GenBank mRNA | AF221521.1 | - | 2 - 3 | 138 | 60 | |
| AK023219.1 | - | 2 - 3 | 130 | 60 | ||
| AK300544.1 | - | 2 - 3 | 153 | 60 | ||
| AK315551.1 | - | 2 - 3 | 170 | 60 | ||
| BC016852.1 | - | 2 - 3 | 117 | 3900461 | 60 | |
| HY108652.1 | - | 2 - 3 | 189 | 60 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003298.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01050920.hg.1 | TC01003298.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050920.hg.1 | ENST00000351146.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050920.hg.1 | ENST00000368460.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050920.hg.1 | ENST00000368463.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050920.hg.1 | ENST00000368465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050920.hg.1 | ENST00000498553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050920.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219973.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219973.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219973.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219973.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219973.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219973.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219973.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219973.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219973.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219973.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219973.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219973.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219973.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219973.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219973.hg.1 | TC0100015932.hg.1 | Clariom D TC |

PBXIP1
Genomic Map