Assay Details
| Gene Symbol: PBXIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317734.1 | NP_001304663.1 | 7 - 8 | 775 | 58 | |
| NM_001317735.1 | NP_001304664.1 | 5 - 6 | 444 | 58 | ||
| NM_020524.3 | NP_065385.2 | 8 - 9 | 864 | 58 | ||
| GenBank mRNA | AF221521.1 | - | 8 - 9 | 813 | 58 | |
| AK022497.1 | - | 7 - 8 | 734 | 58 | ||
| AK023219.1 | - | 8 - 9 | 805 | 58 | ||
| AK300987.1 | - | 4 - 5 | 153 | 58 | ||
| AK303413.1 | - | 5 - 6 | 388 | 58 | ||
| AK303863.1 | - | 5 - 6 | 289 | 58 | ||
| AK315551.1 | - | 8 - 9 | 845 | 58 | ||
| BC016852.1 | - | 8 - 9 | 792 | 3900461 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01050906.hg.1 | TC01003298.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050906.hg.1 | ENST00000351146.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050906.hg.1 | ENST00000368463.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050906.hg.1 | ENST00000368465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050906.hg.1 | ENST00000539880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050906.hg.1 | ENST00000542459.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050906.hg.1 | ENST00000543593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050906.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000368465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000490230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000493133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | ENST00000498553.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015932.hg.1 | NM_020524.3 | Refseq |

PBXIP1
Genomic Map