Assay Details
| Gene Symbol: FAM63A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163260.1 | NP_001156732.1 | 1 - 2 | 481 | 93 | |
| GenBank mRNA | AK002142.1 | - | 1 - 2 | 331 | 93 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003216.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK002142.1,NM_001163260.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01049435.hg.1 | TC01003216.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049435.hg.1 | ENST00000312210.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049435.hg.1 | ENST00000361936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049435.hg.1 | ENST00000470877.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049435.hg.1 | ENST00000493834.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049435.hg.1 | NM_001040217.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049435.hg.1 | NM_001163259.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049435.hg.1 | NM_001163260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049435.hg.1 | NM_018379.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049429.hg.1 | TC01003216.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049429.hg.1 | ENST00000312210.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049429.hg.1 | ENST00000361738.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049429.hg.1 | ENST00000361936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049429.hg.1 | ENST00000493834.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049429.hg.1 | NM_001040217.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049429.hg.1 | NM_001163258.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049429.hg.1 | NM_001163259.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049429.hg.1 | NM_001163260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049429.hg.1 | NM_018379.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049436.hg.1 | TC01003216.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049436.hg.1 | ENST00000312210.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049436.hg.1 | ENST00000361936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049436.hg.1 | ENST00000470877.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049436.hg.1 | ENST00000493834.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049436.hg.1 | NM_001040217.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049436.hg.1 | NM_001163259.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049436.hg.1 | NM_001163260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01049436.hg.1 | NM_018379.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218196.hg.1 | ENST00000312210.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218196.hg.1 | ENST00000361936.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218196.hg.1 | ENST00000470877.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218196.hg.1 | ENST00000493834.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218196.hg.1 | NM_001040217.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218196.hg.1 | NM_001163259.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218196.hg.1 | NM_001163260.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218196.hg.1 | NM_018379.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218196.hg.1 | ENST00000312210.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218196.hg.1 | ENST00000361936.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218196.hg.1 | ENST00000470877.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218196.hg.1 | ENST00000493834.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218196.hg.1 | NM_001040217.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218196.hg.1 | NM_001163259.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218196.hg.1 | NM_001163260.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218196.hg.1 | NM_018379.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218196.hg.1 | TC0100015764.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | ENST00000312210.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | ENST00000361738.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | ENST00000361936.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | ENST00000493834.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | ENST00000622754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | NM_001040217.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | NM_001163258.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | NM_001163259.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | NM_001163260.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | NM_018379.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | ENST00000312210.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | ENST00000361738.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | ENST00000361936.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | ENST00000493834.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | ENST00000622754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | NM_001040217.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | NM_001163258.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | NM_001163259.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | NM_001163260.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | NM_018379.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218186.hg.1 | TC0100015764.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100218195.hg.1 | ENST00000312210.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218195.hg.1 | ENST00000361936.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218195.hg.1 | ENST00000470877.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218195.hg.1 | ENST00000493834.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218195.hg.1 | ENST00000622754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218195.hg.1 | NM_001040217.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218195.hg.1 | NM_001163259.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218195.hg.1 | NM_001163260.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218195.hg.1 | NM_018379.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218195.hg.1 | ENST00000312210.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218195.hg.1 | ENST00000361936.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218195.hg.1 | ENST00000470877.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218195.hg.1 | ENST00000493834.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218195.hg.1 | ENST00000622754.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100218195.hg.1 | NM_001040217.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218195.hg.1 | NM_001163259.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218195.hg.1 | NM_001163260.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218195.hg.1 | NM_018379.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100218195.hg.1 | TC0100015764.hg.1 | Clariom D TC |

FAM63A
Genomic Map