Assay Details
| Gene Symbol: CACNA1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129833.1 | NP_001123305.1 | 30 - 31 | 4227 | 72 | |
| NM_001129841.1 | NP_001123313.1 | 30 - 31 | 4227 | 72 | ||
| NM_001129842.1 | NP_001123314.1 | 30 - 31 | 4227 | 72 | ||
| NM_001129844.1 | NP_001123316.1 | 30 - 31 | 4218 | 72 | ||
| NM_001167623.1 | NP_001161095.1 | 30 - 31 | 4227 | 72 | ||
| NM_001167624.2 | NP_001161096.2 | 30 - 31 | 4192 | 72 | ||
| XM_011521020.2 | XP_011519322.1 | 31 - 32 | 4356 | 72 | ||
| XM_017019934.1 | XP_016875423.1 | 32 - 33 | 5543 | 72 | ||
| XM_017019937.1 | XP_016875426.1 | 31 - 32 | 5468 | 72 | ||
| XM_017019944.1 | XP_016875433.1 | 31 - 32 | 5468 | 72 | ||
| XM_017019946.1 | XP_016875435.1 | 31 - 32 | 5468 | 72 | ||
| XM_017019948.1 | XP_016875437.1 | 31 - 32 | 5468 | 72 | ||
| XM_017019949.1 | XP_016875438.1 | 31 - 32 | 5468 | 72 | ||
| XM_017019951.1 | XP_016875440.1 | 31 - 32 | 5459 | 72 | ||
| XM_017019953.1 | XP_016875442.1 | 30 - 31 | 4281 | 72 | ||
| XM_017019954.1 | XP_016875443.1 | 30 - 31 | 4281 | 72 | ||
| GenBank mRNA | AY830712.1 | - | 30 - 31 | 3914 | 72 | |
| BC146846.1 | - | 30 - 31 | 4033 | 9056774 | 72 | |
| L29534.1 | - | 29 - 30 | 3840 | 72 | ||
| L29536.1 | - | 29 - 30 | 3840 | 72 | ||
| M57972.1 | - | 4 - 5 | 431 | 72 | ||
| Z34809.1 | - | 30 - 31 | 3914 | 72 | ||
| Z34816.1 | - | 30 - 31 | 3914 | 72 | ||
More Information
Set Membership: |
|

CACNA1C
Genomic Map